Wide Mouth Rounds, High-Density Polyethylene
Supplier: Qorpak
PLA-03309 PLA-06145 PLC-06146 PLC-03580 PLC-03606 PLC-03594 PLC-03579 PLC-03555 PLC-03533 PLA-03321 PLC-03536 PLC-03583 PLA-03312 PLA-03317 PLA-03322 PLC-03597 PLA-10061 PLC-03619 PLC-03645 PLC-03547 PLC-03618 PLA-03311 PLC-03564 PLA-03315 PLA-03318 PLC-03637 PLC-03639 PLC-03526 PLC-03609 PLC-03600 PLC-03610
11311-234CS 339.93 CAD
11311-234 11311-364 11311-620 15000-400 15000-402 15000-404 16054-274 16081-035 16081-041 16081-044 16081-060 16085-238 16087-150 16087-310 16125-812 16151-820 16218-146 16218-160 16218-161 16218-167 36319-302 36319-341 36319-342 36319-359 89009-336 89085-344 93001-750 16151-804 16151-818 16151-834 16151-858 36319-300 89204-900 89204-902 16081-059 16218-139 11311-393 30617-144 30618-710 30618-712 76076-773 76487-178 76487-180 76539-380 76539-382
Wide Mouth Rounds, High-Density Polyethylene
Bottles Roller Bottles
Natural HDPE wide mouth round bottles are ideal for general lab use with solids and powders.
  • Available in multiple sizes, from 1 to 128 oz.
  • Available with a variety of caps or no caps
  • Bulk or convenience packs offered
These HDPE bottles provide an excellent moisture barrier and are impact resistant. They are ideal for high-speed filling, demanding pharmaceutical specifications, and the rough handling of interstate shipping.
Ordering information: Bottles 36319-300, 36319-302, 36319-359, 16125-812, 16125-820, and 93001-750 are supplied pre-cleanedMore Product Information