Nalgene® Large Polypropylene Copolymer Bottles, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2204-0005 2204-0010 2204-0020 2203-0005 2203-0010 2203-0020
16062-142CS 273.02 CAD
16062-142 16062-164 16062-186 16067-146 16067-168 16067-180
Nalgene® Large Polypropylene Copolymer Bottles, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Thermo Scientific Nalgene® Large Narrow Mouth Bottles feature excellent chemical resistance to most acids, bases, and alcohols, making them ideal for storing chemicals and standards as well as collecting and storing distilled water.
The 3.8 L and 8 L sizes feature a built-in shoulder loop for attaching an identification tag. All bottles are supplied with polypropylene screw closures. The 8 L bottle includes a white PP closure with a TPE gasket.
Avavilable in natural and amber finishes. Amber bottles comply with U.S. Pharmacopoeia 23, 661 requirements for maximum allowable lightMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable