Boston Bottles, Round, Low-Density Polyethylene, Narrow Mouth, Qorpak®
Supplier: Qorpak
PLC-03661 PLC-03668 PLC-03674 PLC-03683 PLA-03332 PLA-03337 PLA-03331 PLA-03333 PLC-03677
16054-002CS 309.26 CAD
16054-002 16054-003 16054-004 16054-005 16054-006 16087-134 16218-144 16807-132 16218-143 75856-978 16085-001 76311-220
Boston Bottles, Round, Low-Density Polyethylene, Narrow Mouth, Qorpak®
Bottles Plastic Bottles
Natural LDPE, round bottles are the perfect solution for liquid sample handling in a squeezable bottle.
These bottles are virtually unbreakable and more resistant to stress cracking than HDPE bottles. They retain their impact strength at very low temperatures.