Tychem® Hoods, Bullard®
Supplier: Bullard
20TJ 20SIC 20SICH 20SICN 20TICN 20TICS 20TJN RT1 20TICH RT3T 20TICSN
16005-040PK 589.02 CAD
16005-040 16006-040 16006-042 16006-044 16006-048 16006-050 16006-054 16006-300 16005-038 89093-555 89235-851 16006-053
Tychem® Hoods, Bullard®
Respirators Respiratory Systems
Tychem® hoods provide a variety of simple, effective, and economical options for respiratory protection.
  • Simple, effective, and economical options for respiratory protection
  • Covers entire head, face, and neck to provide splash and overspray protection
  • Neck cuff design maintains positive pressure inside the hood by inflating the inner collar to provide a snug, comfortable fit and continuous airflow
  • Wide-angle lens offers large field of vision while accommodating beards, prescription eyeglasses, or safety glasses
  • Lighter and more comfortable than tight-
More Product Information