Precision Stat® Class 100 Centerfold Film, PPC Flexible Packaging
Supplier: Fisher Container
2N600720F STCK0040-001
16003-918EA 2648.05 CAD
16003-918 21915-012
Precision Stat® Class 100 Centerfold Film, PPC Flexible Packaging
Prefabricated Rooms
This polyethylene film features an inner surface suitable for use in ISO Class 5 (FED-STD 209E Class 100/M3.5) cleanroom conditions
It is clear, with a special amine-free, antistatic formulation. To prevent contamination, film is shipped on plastic cores in individual cartons.
Order Now
SPECIFICATIONS
Environmentally Preferable