Detachable Thermocouples, Process Reactors, Type T and K, Chemglass
Supplier: Chemglass
CG-3498-34 CG-3498-35 CG-3498-120 CG-3498-100 CG-3498-101 CG-3498-102 CG-3498-104 CG-3498-105 CG-3498-106 CG-3498-107 CG-3498-108 CG-3498-109 CG-3498-110 CG-3498-111 CG-3498-200 CG-3498-201 CG-3498-202 CG-3498-203 CG-3498-204 CG-3498-205 CG-3498-206 CG-3498-207 CG-3498-208 CG-3498-209 CG-3498-300 CG-3498-301 CG-3498-302 CG-3498-303 CG-3498-304 CG-3498-305 CG-3498-306 CG-3498-307 CG-3498-309 CG-3498-30 CG-3498-32 CG-3498-116 CG-3498-118
16003-438EA 352.9 CAD
16003-438 16003-440 89166-650 16003-354 16003-356 16003-358 16003-360 16003-362 16003-364 16003-366 16003-368 16003-370 16003-372 16003-374 16003-376 16003-382 16003-384 16003-386 16003-388 16003-390 16003-392 16003-394 16003-396 16003-398 16003-400 16003-410 16003-412 16003-414 16003-416 16003-418 16003-420 16003-422 16003-424 16003-426 16003-428 47734-132 47734-136 89030-034 89030-038
Detachable Thermocouples, Process Reactors, Type T and K, Chemglass
Temperature Probes
The detachable thermocouples are for direct insertion in the reactors using a CG-1048-A or CG-1042-E thermocouple adapter, while the 4” long Hastelloy C-276 style is for use with the CG-1971 baffles and adapters.
  • Thermocouples for use with Chemglass process reactors
  • Available in both “T” (blue connections) and “K” (yellow connections)
Detachable type thermocouples have a standard male plug (OST) and require a coiled extension cord for connection to the CG-3498 temperature monitor
Note: The 1/2” dia. probes have a transition to 6” of PTFE leads andMore Product Information