XRF Thin-Film Sample Supports, Chemplex®
Supplier: Chemplex
476 436 425 426 416 256 250 156 150 106 100 090 475
15566-008PK 415.22 CAD
15566-008 15566-020 15566-022 15566-024 15566-026 15566-028 15566-030 15566-034 15566-036 15566-038 15566-040 15566-042 15566-044 15566-048 15566-050 15566-052 15566-058 15566-010
XRF Thin-Film Sample Supports, Chemplex®
Sample Containers XRF Sample Cups
For retaining liquid, powdered, slurry or solid specimens in XRF Sample Cups for wavelength (WDXRF) and energy dispersive (EDXRF) spectrochemical analysis.
  • Chemplex thin-film sample support substances are manufactured to very close tolerances (1 to 2% within a single package)
Choose from Prolene, ultra-polyester, polypropylene, Mylar, polycarbonate or polyimide support materials based on their ability to safely retain specimen during preparation and analysis stages, permit maximum analyte-line transmittance, and provide statistically reproducibleMore Product Information