Econo™ Animal Cages, Maryland Plastics
Supplier: Maryland Plastics
E0250 E0270 E0494B E0594B E0150 E0495B E0550 E0370 E1470 E1450 E0211
10717-715CS 812.88 CAD
10717-715 10717-702 10717-704 10718-066 10718-068 10718-094 10718-096 10718-202 10718-204 10718-354 10718-356 15431-360 73000-202 73000-214 73000-219
Econo™ Animal Cages, Maryland Plastics
Animal Supplies Animal Cages
Econo reusable cages are designed for the housing and/or breeding of rats, mice, hamsters, gerbils, and guinea pigs.
Durable polycarbonate cages are clear and have extremely high impact strength. They are highly resistant to chemicals and solvents, and practically unbreakable. In addition, they can withstand autoclaving up to 121 °C (250 °F). Economical polypropylene cages are translucent with good impact strength, and the smooth surface allows for easy cleaning. They are highly resistant to chemicals and solvents and can withstand temperatures up to 110 °C (230 °F).
The disposableMore Product Information