Fuchs-Rosenthal Counting Chamber, Hausser Scientific
Supplier: Hausser
3720
15170-230EA 977.12 CAD
15170-230
Fuchs-Rosenthal Counting Chamber, Hausser Scientific
Slides Counting Chambers
Hemacytometer for counting cells in cerebrospinal fluid, etc
Cell depth: 0.2 mm. Fuchs-Rosenthal ruling with sixteen 1mm squares each subdivided into 16 smaller squares. One piece construction. Supplied with two cover glasses. Without case.