Heavy-Duty Storage Cabinets, SECURALL®
Supplier: Securall Cabinets
SS142 SS172 SS184
15001-716EA 3023.19 CAD
15001-716 15001-718 15001-720
Heavy-Duty Storage Cabinets, SECURALL®
Furniture Storage Cabinets
Constructed of 18-gauge galvanized steel, these heavy-duty storage cabinets feature two double-wall doors and a locking, flush-mount handle to prevent unauthorized entry.
  • Custom interiors are available to fit the specific needs
  • 18-gauge all-welded extra strength construction throughout
Cabinets have the SUR-LOC™ adjustable shelving system and can store up to 226.8 kg (500 lbs.) per shelf. Additional features include three-point self-latching button closure and reinforced side walls.
Color: Gray
Ordering information: Beige cabinets are alsoMore Product Information