Uvex Genesis® Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
S3203 S3240 S3241X S3244 S3200 S3240 S3200HS S3201HS
14216-001EA 59.65 CAD
14216-001 14227-817 CA14216-000 CA14216-002 CA14227-815 CA14231-228 CA15001-568 CA15001-570 CA15001-576 CA15001-578 CA15001-582 CA15001-610 CA15001-612 CA15001-616 CA15001-620 CA15001-622 CA94023-916 15001-568 15001-610 10790-624 10790-626 CA14227-817
Uvex Genesis® Protective Eyewear, Honeywell Safety
Safety Glasses
Designed for comfort and compliance, Uvex Genesis® eyewear combines a lightweight, resilient multi-material frame with a soft elastomer brow guard for unsurpassed impact resistance.
  • Dual 9-base wraparound lens provides uncompromised peripheral vision and protection Elastomer brow-guard diffuses and deflects impact
  • Temple length and lens inclination are adjustable
  • Soft fingers conform to any nasal profile
  • Frames guaranteed for life
The dual 9-base wraparound polycarbonate lens provides uninterrupted peripheral coverage and visibility.
Clear lensesMore Product Information