Anticon® Polywipe StandardWeight, Heatsealed, Unlaundered, Contec
Supplier: Contec
495356-009
14900-518CS 645.22 CAD
14900-518
Anticon® Polywipe StandardWeight, Heatsealed, Unlaundered, Contec
Wipes Disposable Wipes and Towels Wipes for General Applications
Anticon® Polywipe StandardWeight, heatsealed automotive wipes incorporate proprietary edge sealing technology with 100% continuous filament polyester chemically modified for superior cleanliness and sorbency.
  • High sorbency capacity
  • Very low particle generation
  • Can be relaundered multiple times
  • Stronger and cleaner than nonwoven wipes
Approved by major Automotive OEMs.
Ideal for sealer clean-up. Use for general maintenance tasks where cleanliness is required. Superb pick-up and holding capabilities.
The thermally sealed, non-abrasive edgeMore Product Information