Disposable Serological Pipettes, Plastic, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
56900-25210 56800-11100 56900-21100 56900-11100 56900-5110 56900-10110 56900-50510 56900-1110
14672-900CS 797.82 CAD
14672-900 14672-904 14672-916 14672-918 14672-920 14672-922 53384-451 14672-914
Disposable Serological Pipettes, Plastic, Kimble Chase
Pipettes Serological Pipettes
These accurate, plastic pipettes are sterile, plugged, and ready for use.
These color coded pipettes feature a black printed scale with reverse and negative graduations to increase the versatility of use. Designed from ASTM Specification E934, and ASTM Specification E1380 for color coding.
Pipettes feature a uniform molded tip and top ends with cotton plugging. They are calibrated to deliver to the tip, with two rings near the top denoting that the small amount remaining after free delivery must be blown out to obtain total rated capacity.
Ordering information: Packaged individuallyMore Product Information