Samco™ Extra Long Transfer Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
262-1S 263 263-1S 262-20S 263-20S 262-10S
14670-395CS 2314.1 CAD
14670-395 14670-398 14670-399 14670-400 14670-401 14670-405 73990-116
Samco™ Extra Long Transfer Pipettes, Thermo Scientific
Pipettes Pasteur/Transfer Pipettes
Perform safe and easy sampling from large containers using Thermo Scientific™ Samco™ Extra-Long Transfer Pipettes.
Perfect for tissue culture applications, these pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, autoclaving and prevents cross-contamination. Pipettes are made of low-density polyethylene.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Environmental certification ISO14001 (Certificate Registration No. EMS 752603) ISO14001 (Certificate Registration No. EMS 752603) ISO14001 (Certificate Registration No. EMS 752603)