Titan3™ PTFE (Hydrophobic) Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
44504-NP 42225-NP 44513-NP 44525-NP 41025-NP 42204-NP 42225-PC
14232-522PK 271.49 CAD
14232-522 14232-746 14232-749 14232-752 14232-882 14234-952 14235-364
Titan3™ PTFE (Hydrophobic) Syringe Filters, Thermo Scientific
Filters Syringe Filters
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.
  • Excellent chemical resistance for use with organic matrices
  • Naturally hydrophobic membrane provides excellent flow rates and high loading capacities
  • Exceptional temperature stability, able to filter solutions at temperatures up to 110 °C
  • Organic solvent recommended
Available in sizes and membrane types to meet any application need, Thermo Scientific™ Titan3™ HydrophobicMore Product Information