Finntip® Filtered Pipette Tips, Thermo Scientific
Supplier: Thermo Fisher Scientific
94052350 94052310 94052020 94052410 94052320 94052300 94052200 94052100 94052000 94052550 94052160 94052060 94052430 94052150 94052800
11310-904PK 461.63 CAD
11310-904 12777-872 14222-044 14233-402 46620-286 53515-262 53515-898 53515-900 53515-908 53515-910 53515-984 11388-542 14224-026 14233-404 46600-006 76637-686
Finntip® Filtered Pipette Tips, Thermo Scientific
Pipette Tips
Finntip Filter Pipette Tips provide superior protection against contamination.
  • Sterilized with irradiation
  • Certified free from DNA, DNase, RNase and endotoxin
Finntip filter pipette tips are available in racks with an assortment of volumes from 0.2 to 10 μl. Made from an inert non-sealing polypropylene matrix, the filter is effective at eliminating contamination from the pipetter to the sample, thereby ensuring the integrity of any pipetting function.
For 14233-402, CS of 10, there are 10 racks in a case, and each rack has 96 tips.
These sterileMore Product Information