Tapered Tissue Grinders, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
358133 358103 358107 358115 358137 358141 358145
14231-524CS 293.4 CAD
14231-524 14231-518 14231-522 14231-542 14231-544 14231-546 14231-548 14231-550
Tapered Tissue Grinders, Wheaton
Homogenizers Tissue Grinders
Grinders are available with glass or fluoropolymer resin pestles
Grinding chamber is made of low extractable borosilicate glass. The conical tip of the grinding chamber allows initial reduction, followed by passage through the cylindrical section for the final homogenization step. When pestle is “bottomed” within the grinding chamber, the clearance is a uniform 0.102–0.152mm (0.004–0.006").
Glass pestles are ideal for the homogenization of heart, muscle, lung, skin, and plant tissue. Fluoropolymer resin pestles are ideal for use with soft tissues such as brain and feature a stainlessMore Product Information