WHEATON® Clear Specimen Bottles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
221624
14227-242CS 220.01 CAD
14227-242
WHEATON® Clear Specimen Bottles, DWK Life Sciences
Bottles Glass Bottles
WHEATON® clear specimen bottles are made from high-quality USP Type III clear soda-lime glass.
  • 125 ml capacity
  • Clear finish
Delivery information: Supplied with rubber lined cap.