HT8-PCB GC Capillary Column, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
054690 054236
14226-014EA 898.02 CAD
14226-014 10811-186
HT8-PCB GC Capillary Column, Trajan Scientific and Medical
Chromatography Columns
More than five decades of innovative phase technologies and unique fused silica production capabilities, together provide end-to-end separation solutions for all applications.
  • 8% Phenyl Polycarborane-siloxane
  • Preferred column for polychlorinated biphenyl (PCB) compounds
  • Separates PCB’s on ortho ring substitution as well as boiling point
  • Low bleed
For use with PCB congener analyses, nitro-substituted aromatics, polynuclear aromatic hydrocarbons and pesticides/herbicides.
Certifications: RoHS compliant per Directive 2011/65/EU.