Lifesaver® Ceiling-Mount Emergency Shower, Speakman®
Supplier: Speakman
SE-236-PR
14224-248EA 2327.02 CAD
14224-248
Lifesaver® Ceiling-Mount Emergency Shower, Speakman®
First Aid Supplies Showers
Lifesaver® emergency shower features a concealed ceiling mounted impeller action chrome plated shower head with a horizontal supply.
Shower is activated with a 2.5 cm (1") stay-open ball valve operated by a high visibility yellow pull rod.
Certifications: Meets ANSI Z358.1 standard.