SP Wilmad-LabGlass Volumetric Serialized Flasks with Stopper, Class A, SP Industries
Supplier: SP Industries
LG-8112-118
14201-984EA 345.72 CAD
14201-984 14201-992 14202-250 14206-328 14207-028 14209-960 14210-520 14203-602 14206-356
SP Wilmad-LabGlass Volumetric Serialized Flasks with Stopper, Class A, SP Industries
Flasks Volumetric Flasks
Glass quality, calibration lines, inscription and appearance of these borosilicate glass flasks are maintained according to the former NBS circular 602.
  • They have a marking spot in addition to a serial number
  • Calibrated “to contain” (TC) at 20 °C
  • Supplied with stopper
Order Now
SPECIFICATIONS
Environmentally Preferable