SP Wilmad-LabGlass Buchner Filter Funnels with Fritted Disc and Vacuum Adapter, SP Industries
Supplier: SP Industries
LG-7120-125
14202-636EA 275.66 CAD
14202-636 14203-360 14203-364 14203-464 14203-466 14204-040 14204-866 14205-760 14205-762 14205-766 14205-768 14206-614 14206-616 14206-618 14207-806 14207-810 14209-246 14209-248 14209-290 14209-292 14209-294 14209-684 14209-686 14209-688 14209-998 14210-000 14210-478 14210-482
SP Wilmad-LabGlass Buchner Filter Funnels with Fritted Disc and Vacuum Adapter, SP Industries
Funnels Filtration Funnels Büchner Funnels
These Buchner filter funnels with vacuum adapter feature a full length Standard Taper inner joint sealed to the stem with side hose connection just above the grinding.
  • Made of borosilicate glass
With sealed-in fritted glass disc, they are available in coarse (C) and medium (M) porosities.
Recommended for use with LG-7820 heavy wall filter flasks.