SP Wilmad-LabGlass Spherical Reaction Vessels, Conical Flange, SP Industries
Supplier: SP Industries
LG-8020-108
14203-920EA 1606.88 CAD
14203-920 14205-048 14205-142 14205-144 14205-238 14205-486 14207-542 14208-016 14209-620
SP Wilmad-LabGlass Spherical Reaction Vessels, Conical Flange, SP Industries
Reaction Vessels Reaction Vessels, Flask Shape
These reaction vessels are made of borosilicate glass and have a spherical body
Upper end of neck is tooled to obtain a thickened portion and flat top. Top is finely ground for a leak-proof fit with lids LG-8030 to LG-8050, ML-1285, and ML-1286.