SP Wilmad-LabGlass Volatile Oil Distilling Apparatus for Oils Lighter than Water, SP Industries
Supplier: SP Industries
LG-6655-100
14205-246EA 1900.45 CAD
14205-246 14208-166 14208-494
SP Wilmad-LabGlass Volatile Oil Distilling Apparatus for Oils Lighter than Water, SP Industries
Distillation Components Distillation Sets
Designed for oils lighter than water, this distilling apparatus is made of borosilicate glass and consists of a Clevenger-type trap with a Standard Taper 24/40 outer joint on top and same size inner joint the sidearm
A 250mL long neck round bottom flask (LG-7221) and a cold finger condenser with a Standard Taper 24/40 inner joint complete the assembly.