SP Wilmad-LabGlass Graduated Cylindrical Addition Funnels with Pressure Equalizing Sidearm, SP Industries
Supplier: SP Industries
LG-8271T-104
14204-882EA 1496.35 CAD
14204-882 14205-130 14207-460 14207-462 14207-464 14207-828 14208-208 14208-212 14208-642 14208-896 14209-494
SP Wilmad-LabGlass Graduated Cylindrical Addition Funnels with Pressure Equalizing Sidearm, SP Industries
Funnels Filtration Funnels Filter Funnels
Featuring graduations with a double scale from the PTFE stopcock, these borosilicate glass cylindrical funnels have a pressure equalizing sidearm
Standard Taper outer joint at the top is the same size as the lower Standard Taper inner joint.
Order Now
SPECIFICATIONS
Environmentally Preferable