SP Wilmad-LabGlass Laboratory Grade Automatic Burettes with PTFE Plug, SP Industries
Supplier: SP Industries
LG-4241T-102 LG-4241T-100
14203-812EA 1128.74 CAD
14203-812 14205-414 14207-608 14208-260 14208-646 14208-928
SP Wilmad-LabGlass Laboratory Grade Automatic Burettes with PTFE Plug, SP Industries
Burettes
This inexpensive portable titration set features a 2mm straight stopcock with PTFE plug
The borosilicate glass burette is filled by pressure supplied by the rubber bulb. A small hole in the filling tube at the "O" line permits setting of the liquid level and excess titrant is returned to the bottle. Complete Set furnished with burette, bottle, rubber hose, rubber stopper, and rubber bulb.