SP Wilmad-LabGlass Arsine Generator, Improved, SP Industries
Supplier: SP Industries
LG-6962-100
14203-134EA 432.67 CAD
14203-134
SP Wilmad-LabGlass Arsine Generator, Improved, SP Industries
Arsine Generators
For arsine analysis using the diethyldithocarbamate colorimetric method, this improved Air-Tite glassware arsine generator is made of borosilicate glass and features a one piece scrubber/absorber tube.
  • Design of apparatus requires less space and is easier to clean than standard ball and socket design
  • Meets ACS/USP and EPA specifications
  • Glass beads size 4mm O.D. (LG-6750-102) may be added to the absorber tube to enhance gas mixing
Ordering information: Complete unit is supplied with scrubber/absorber tube, a 125mL Erlenmeyer flask (LG-7750-122), and aMore Product Information