SP Wilmad-LabGlass Metal Clamps, SP Industries
Supplier: SP Industries
LG-1006-104
14007-390EA 22.45 CAD
14007-390 14007-392 14007-394 14007-396
SP Wilmad-LabGlass Metal Clamps, SP Industries
Clamps General Purpose Clamps
These clamps hold Standard Taper joints in place securely under moderate pressure.
Made of metal, they are coated with silicone lacquer to avoid scratching glass surfaces. They can withstand temperatures up to 200°C, and are suitable where higher temperatures are used or cracking occurs in the plastic equivalent.