SP Wilmad-LabGlass Thin-Walled Precision NMR Sample Tubes, 5 mm, SP Industries
Supplier: SP Industries
528-PP-7-5 528-PP-8-5 541-PP-7-5
13501-500PK 287.47 CAD
13501-500 13501-502 13501-504 13501-506 13501-508 13501-510 13501-512 13501-516 13501-518 13501-520 13501-524 13501-526 14218-716 14218-726 14218-740 14218-956 14218-962 14218-976 14218-986 14219-000 14219-002 14219-004 14219-006 13501-522
SP Wilmad-LabGlass Thin-Walled Precision NMR Sample Tubes, 5 mm, SP Industries
Tubes NMR Tubes
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming.
  • Inner surface is resistant to strong acids and bases at ambient temperature
  • Can be operated safely at temperatures up to 230 °C, and within a temperature step of 120 °C
  • Rated as hydrolytic class 1
  • Made from borosilicate glass that meets the requirement of Type 1 Class A glass from ASTM E438
  • 100% inspection with multiple NIST traceable gauges and optical surface defect checks
  • Note: Caps are not recommended for use with samples dissolved in chloroform-d or
More Product Information