SP Bel-Art Cleanware™ Bottle Brush, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F17068-0000
13196-072EA 145.22 CAD
13196-072
SP Bel-Art Cleanware™ Bottle Brush, Bel-Art Products, a part of SP
Brushes
Uniquely designed, swiveling pivot head enables thorough cleaning of bottles and flasks including difficult curves and corners.
  • Natural bristles set in wood will not scratch glass
  • Brush head is slightly convex
  • Brush head is attached to hardwood handle
The swivel head fits through a 29/42 standard taper neck, or any non-tapered neck with a diameter of 24 mm (1") or larger.