Tax-Fre® Dry Mops, Contec®
Supplier: Contec
UF211 TF-1017QF TF-1228
46610-130EA 160.52 CAD
46610-130 89129-434 94052-810 12777-950
Tax-Fre® Dry Mops, Contec®
Mops
Designed for dry mopping floors and walls, the Tax-Fre® Dry Mop utilizes the same low-residue material as Tax-Fre® wipes.
  • Utilizing a low-residue pressure-sensitive material, this dry floor mop provides superior pick-up in areas where wet mopping is not an option
  • Will not scratch or damage sensitive surfaces
  • Dramatically reduces clean-up time
  • Ideal for use when liquids are not recommended
The 23×5" (58.4×12.7 cm) trapezoid-shaped swivel head attached to a lightweight 5' (1.5 m) handle enables the operator to clean in hard to each areas. All hardwareMore Product Information