OIML Class E2 Precision Weights, Troemner
Supplier: Troemner
80780284 80780285 80780286 80780287 80780288 80780289 80780290 80780291 80780292 80780293 80780294 80780295 80780296 80780297 80780298 80780299 80780300 80780301 80780304 80780303 80780302
12766-154EA 369.81 CAD
12766-154 12766-156 12766-158 12766-160 12766-162 12766-164 12766-166 12766-168 12766-170 12766-172 12766-174 12766-176 12766-178 12766-180 12766-182 12766-184 12766-186 12766-188 12766-190 12766-192 12766-194
OIML Class E2 Precision Weights, Troemner
Weights Calibration Weights
Troemner OIML Calibration Weights conform to the standards of the Organisation Internationale de Metrologie Legale International Recommendation (OIML R 111)
Weights 1 g and above are one-piece construction. Troemner OIML Calibration Weights 1 g and larger are cylindrical in shape with a tapered neck for easy gripping. They are perfectly flat on top to stack easily on center. Weight bottoms are slightly recessed to expose the smallest possible area to wear.
They are manufactured from the finest stainless steels available including Troemner Alloy 8 Stainless Steel that has lowMore Product Information