Electronic Balance Calibration Weights, Ultra Class and ANSI/ASTM Classes 1 and 4, Troemner
Supplier: Troemner
80781007 30391488 80781005 80781008 30391520 30391537 30391519 80781006 30391517 80781003 80781004 30391515 80781001 30391481 80781002 30391513 30391533 30391512 80780027 80850275 30391510 80780024 80850272 30391508 80780021 80850269 30391504 80780026 80850274 30391503 30391471 30391528 30391502 80850271 30391468 30391526 30391499 80850268 30391498 80780028 80850276 30391494 80780025 80850273 30391493 30391461 30391522 30391492 80861108 80861110 80861109 30391139 30391138 30391136 30391170 30391194 30391169 30391193 30391134 30391189 30391165 30391132 30391131 30391187 30391163 30391186 30391162 30391129 30391184 30391160 30391158 80780143 80780144 30391156 80780141 80780142 30391154 80780139 80780140 30391153 80780137 80780138 30391152 80780135 80780136 30391151 30391118 30391179 30391149 80780133 80780134 30391148 30391175 30391144 30391112 30391174 30391143 30391111 30391173 30391142 30390301 30390300 30390298 30390296 30390294 30390293 30390291 30390289 30390287 30390285 30390284 30390283 30390282 30390281 30390280 30390279 30390275 30390274 30390273 80860833 30390363 30390362 30390360 30390358 30390356 30390355 30390353 30390349 30390347 30390346 30390345 30390344 30390343 30390342 30390341 30390337 30390336 30390335 80860835 30390332 30390331 30390329 30390327 30390325 30390324 30390322 30390320 30390318 30390316 30390315 30390314 30390313 30390312 30390311 30390310 30390306 30390305 30390304 80860834 80861554 80861553 80861552 30391506 80780029 80850277 80780023 30391501 80780020 30391167 30391191 80780145 80780146 30391113 30390351
12620-084EA 5272.49 CAD
12620-084 12620-086 12620-090 12620-092 12620-093 12620-094 12620-095 12620-096 12620-097 12620-159 12620-160 12620-161 12620-217 12620-219 12620-220 12620-221 12620-222 12620-223 12620-239 12620-240 12620-241 12620-363 12620-364 12620-365 12620-421 12620-422 12620-489 12620-523 12620-524 12620-525 12620-545 12620-546 12620-547 12620-580 12620-636 12620-638 12620-639 12620-670 12620-671 12620-771 12620-772 12620-773 12620-793 12620-794 12620-795 12620-818 12620-820 12620-821 12620-835 12620-836 12620-837 12623-096 12623-098 12623-102 12623-103 12623-104 12623-105 12623-106 12623-168 12623-170 12623-171 12623-226 12623-228 12623-230 12623-231 12623-232 12623-233 12623-339 12623-340 12623-341 12623-375 12623-430 12623-432 12623-433 12623-496 12623-498 12623-499 12623-532 12623-534 12623-535 12623-554 12623-556 12623-557 12623-587 12623-588 12623-589 12623-645 12623-646 12623-647 12623-678 12623-680 12623-681 12623-772 12623-773 12623-791 12623-792 12623-793 12623-827 12623-828 12623-829 12765-728 12765-730 12765-732 12765-734 12765-736 12765-738 12765-740 12765-742 12765-744 12765-746 12765-748 12765-750 12765-752 12765-754 12765-756 12765-758 12765-760 12765-762 12765-764 12765-766 12765-768 12765-770 12765-772 12765-774 12765-776 12765-778 12765-780 12765-784 12765-786 12765-788 12765-790 12765-792 12765-794 12765-796 12765-798 12765-800 12765-802 12765-804 12765-806 12765-808 12765-810 12765-812 12765-814 12765-816 12765-818 12765-820 12765-822 12765-824 12765-826 12765-828 12765-830 12765-832 12765-834 12765-836 12765-838 12765-840 12765-842 12765-844 12765-846 12765-848 12765-850 12765-910 12620-423 12620-487 12620-488 12620-578 12620-581 12620-669 12623-107 12623-108 12623-372 12623-374 12623-770 12765-782
Electronic Balance Calibration Weights, Ultra Class and ANSI/ASTM Classes 1 and 4, Troemner
Weights Calibration Weights
Precision-crafted of stainless steel, with a highly polished finish.
Weights 30kg to 5kg are flat on top with a recessed grip handle for easy handling and stacking
Weights 5kg and below supplied in a rugged, precision-fit, high quality polycarbonate case with handling glove.
Weights 5kg to 1kg are cylindrical in shape with a groove near the top to allow for easy handling and gripping. Weights below 1kg are straight cylinders. All are manufactured with adjusting cavities.
Certifications: Where applicable, weights are supplied with a NIST/NVLAP Weight Calibration Certificate or aMore Product Information