Alloy 8 Individual Precision Analytical Weights, Ultra Class and ANSI/ASTM Classes 1 and 4, Troemner
Supplier: Troemner
80781144 80781101 80781103 30391395 80781107 80781109 80781111 30391399 80781115 80781117 80781119 30391403 80781123 80781125 80781127 30391407 80781135 30391411 80781139 80781141 80781102 80781104 30391428 80781108 80781112 30391432 80781116 30391436 80781126 80781128 30391440 80781132 80781134 80781136 30391444 80781140 30391575 30391576 30391577 30391578 30391579 30391580 30391581 30391582 30391584 30391585 30391586 30391587 30391588 30391589 30391590 30391592 30391593 30391594 30391595 80780058 80780064 30391265 80780071 80780059 80780065 30391269 80780072 80780060 80780066 30391273 80780067 30391277 80780074 80780062 80780068 30391281 80780075 80780063 80850282 80850288 30391298 80850293 80850289 30391302 80850284 80850290 30391306 80850295 30391310 80850296 80850292 30391314 80850297 80850287 30391076 30391077 30391078 30391079 30391080 30391081 30391082 30391083 30391084 30391085 30391086 30391087 30391088 30391089 30391090 30391091 30391092 30391094 30391096 30390191 30390190 30390189 30390188 30390187 30390186 30390185 30390184 30390183 30390182 30390180 30390179 30390178 30390177 30390176 30390175 30390174 30390173 30390172 30390257 30390255 30390254 30390253 30390252 30390251 30390250 30390249 30390248 30390247 30390246 30390244 30390243 30390242 30390241 30390240 30390239 30390224 30390223 30390222 30390221 30390220 30390219 30390218 30390217 30390216 30390215 30390214 30390213 30390212 30390211 30390210 30390209 30390208 30390207 30390206 30390205 30390270 30390199 30390264 30390231 30390197 30390262 30390229 30390196 30390261 30390228 30390195 30390260 30390227 30390193 30390258 30390225 30391420 30391453 30391602 30391418 30391451 30391600 30391417 30391450 30391599 30391416 30391598 30391596 30391290 30391323 30391103 30391288 30391321 30391101 30391287 30391320 30391100 30391286 30391319 30391099 30391284 30391097 80781131 80781133 80781110 80781118 80781120 80781124 80781142 30391583 30391591 80780073 80780061 80850283 80850294 80850285 80850291 80850286 30391093 30391095 30390192 30390181 30390256 30390245 30390238 30390237 30391449 80781143 30391317 80780073 80780072 80780071 80780066 80780065 80780064 80780060 80780059 80780058 80780063
12755-556EA 817.91 CAD
12755-556 12764-900 12764-902 12764-904 12764-906 12764-908 12764-910 12764-912 12764-914 12764-916 12764-918 12764-920 12764-922 12764-924 12764-926 12764-928 12764-934 12764-936 12764-938 12764-940 12764-950 12764-951 12764-952 12764-953 12764-955 12764-956 12764-957 12764-960 12764-962 12764-963 12764-964 12764-965 12764-966 12764-967 12764-968 12764-969 12764-975 12764-976 12764-977 12764-978 12764-979 12764-980 12764-981 12764-982 12764-984 12764-985 12764-986 12764-987 12764-988 12764-989 12764-990 12764-992 12764-993 12764-994 12764-995 12765-400 12765-402 12765-404 12765-406 12765-408 12765-410 12765-412 12765-414 12765-416 12765-418 12765-420 12765-426 12765-428 12765-430 12765-432 12765-434 12765-436 12765-438 12765-440 12765-510 12765-511 12765-512 12765-513 12765-515 12765-516 12765-518 12765-519 12765-520 12765-521 12765-524 12765-525 12765-527 12765-528 12765-529 12765-530 12765-535 12765-536 12765-537 12765-538 12765-539 12765-540 12765-541 12765-542 12765-543 12765-544 12765-545 12765-546 12765-547 12765-548 12765-549 12765-550 12765-551 12765-553 12765-555 12765-602 12765-604 12765-606 12765-608 12765-610 12765-612 12765-614 12765-616 12765-618 12765-620 12765-624 12765-626 12765-628 12765-630 12765-632 12765-634 12765-636 12765-638 12765-640 12765-642 12765-646 12765-648 12765-650 12765-652 12765-654 12765-656 12765-658 12765-660 12765-662 12765-664 12765-670 12765-672 12765-674 12765-676 12765-678 12765-680 12765-686 12765-688 12765-690 12765-692 12765-694 12765-696 12765-698 12765-700 12765-702 12765-704 12765-706 12765-708 12765-710 12765-712 12765-714 12765-716 12765-718 12765-720 12765-722 12765-724 12765-726 12766-500 12766-502 12766-504 12766-506 12766-508 12766-510 12766-512 12766-514 12766-516 12766-518 12766-520 12766-522 12766-524 12766-526 12766-528 12766-530 12766-532 12766-534 12766-536 12766-538 12766-540 12766-542 12766-544 12766-546 12766-548 12766-552 12766-558 12766-560 12766-562 12766-564 12766-566 12766-568 12766-570 12766-572 12766-574 12766-576 12766-578 12766-580 12766-582 12766-584 12766-588 12764-930 12764-932 12764-954 12764-958 12764-959 12764-961 12764-970 12764-983 12764-991 12765-422 12765-424 12765-514 12765-517 12765-522 12765-523 12765-526 12765-552 12765-554 12765-600 12765-622 12765-644 12765-668 12765-682 12765-684 12766-550 12766-554 12766-586 73690-036 73690-038 73690-040 73690-042 73690-044 73690-046 73690-048 73690-050 73690-052 73690-054
Alloy 8 Individual Precision Analytical Weights, Ultra Class and ANSI/ASTM Classes 1 and 4, Troemner
Weights Calibration Weights
Individual weights available in Troemner UltraClass (equals or exceeds OIML Class E2 tolerances), and ANSI/ASTM Classes 1 and 4.
Fractional weights 500mg to 5mg are made of stainless steel (30mg weights are aluminum); fractional weights 3mg and below are made of aluminum.
UltraClass weights 1g to 20kg are made from Troemner Alloy 8, which is a specially austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability and resistance to corrosion and handling damage. This exclusive material has a density of 8.0 at 25ºC (controlledMore Product Information