Individual Precision Analytical Weights, Class 2, Troemner
Supplier: Troemner
80858701 80858663 80858636 80858610 80858569 80858530 80858504 80858480 80858442 80858403 80858368 80858328 80858301 80858286 80858272 80858256 80858240 80858226 80858212 80858197 80858181 80858703 80858664 80858637 80858611 80858572 80858532 80858505 80858481 80858443 80858404 80858369 80858329 80858302 80858287 80858273 80858258 80858241 80858227 80858213 80858198 80858182 80858705 80858666 80858638 80858613 80858574 80858534 80858506 80858483 80858445 80858406 80858371 80858331 80858303 80858288 80858274 80858259 80858242 80858228 80858214 80858199 80858183 80858165 80858166 80858167 80858138 80858139 80858124 80858125 80858126 80858112 80858113 80858084 80858085 80858086
12666-050EA 128.34 CAD
12666-050 12666-052 12666-054 12666-056 12666-058 12666-060 12666-062 12666-064 12666-066 12666-068 12666-070 12666-072 12666-074 12666-076 12666-078 12666-080 12666-082 12666-084 12666-086 12666-088 12666-090 12666-100 12666-102 12666-104 12666-106 12666-108 12666-110 12666-112 12666-114 12666-116 12666-118 12666-120 12666-122 12666-124 12666-126 12666-128 12666-130 12666-132 12666-134 12666-136 12666-138 12666-140 12666-150 12666-152 12666-154 12666-156 12666-158 12666-160 12666-162 12666-164 12666-166 12666-168 12666-170 12666-172 12666-174 12666-176 12666-178 12666-180 12666-182 12666-184 12666-186 12666-188 12666-190 14216-748 14216-750 14216-752 14216-760 14216-762 14216-764 14216-772 14216-774 14216-776 14216-786 14216-788 14216-796 14216-798 14216-800
Individual Precision Analytical Weights, Class 2, Troemner
Weights Calibration Weights
Highly polished kilogram and gram weights made of Troemner Alloy 8, which is a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage
Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are aluminum); fractional weights 3 mg and below are made of aluminum.
This exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under anyMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable