Alloy 8 Precision Analytical Weight Sets, Ultra Class and ANSI/ASTM Classes 1 and 4, Troemner
Supplier: Troemner
30391339 30391371 30391552 30391206 30391240 30391053 80781025 80781026 30391549 30391204 30391238 30391050 80781029 80781030 30391554 80781028 80781027 80781023 80781024 30391545 80780111 80850264 30391046 30391208 30391242 30391055 80850265 30391051 30390077 30390075 30390073 30390072 30390061 30390151 30390149 30390147 30390146 30390142 30390135 30390114 30390112 30390110 30390109 30390098 80781021 80781022 30391538 80850263 30390105 30390086 30390160 30390123 30390084 30390158 30390121 30390082 30390156 30390119 30390080 30390154 30390117 30390078 30390152 30391349 30391381 30391563 30391347 30391379 30391561 30391345 30391377 30391559 30391343 30391375 30391557 30391341 30391373 30391555 30391217 30391251 30391064 30391215 30391249 30391062 30391213 30391247 30391060 30391211 30391245 30391058 30391209 30391056 30390008 30390012 30390016 80781031 30390057 80781032 30390045 30390041 30390049 30390010 30390014 30390018 30390053 30390059 30390055 30390047 30390043 30390051 30391550 30390068 80780100 30391039 30390115
12626-245EA 3056.31 CAD
12626-245 12626-246 12626-247 12626-252 12626-253 12626-254 12626-265 12626-266 12626-267 12626-275 12626-276 12626-277 12627-988 12627-989 12627-990 12627-992 12627-993 12627-999 12628-000 12628-001 12665-000 12665-001 12665-002 12665-018 12665-019 12665-020 12665-023 12665-024 12765-852 12765-854 12765-856 12765-858 12765-862 12765-864 12765-866 12765-868 12765-870 12765-872 12765-874 12765-876 12765-878 12765-880 12765-882 12765-884 12765-886 12765-888 12765-890 12765-906 12765-912 12766-590 12766-592 12766-594 12766-596 12766-598 12766-600 12766-602 12766-604 12766-606 12766-608 12766-610 12766-612 12766-614 12766-616 12766-620 12766-622 12766-624 12766-626 12766-628 12766-630 12766-632 12766-634 12766-636 12766-638 12766-640 12766-642 12766-644 12766-646 12766-648 12766-650 12766-652 12766-654 12766-656 12766-658 12766-660 12766-662 12766-664 12766-666 12766-668 12766-670 12766-672 12766-674 12766-678 14216-826 14216-828 14216-830 14216-832 14216-834 14216-836 14216-850 14216-852 14216-854 14216-892 14216-894 14216-896 14216-898 14216-900 14216-902 14216-916 14216-918 14216-920 12627-994 12765-860 12765-904 12765-908 12766-618
Alloy 8 Precision Analytical Weight Sets, Ultra Class and ANSI/ASTM Classes 1 and 4, Troemner
Weights Calibration Weights
Weight set available in Troemner UltraClass (equals or exceeds OIML Class E2 tolerances), and ANSI/ASTM Classes 1 and 4
Fractional weights 500mg to 5mg are made of stainless steel (30mg weights are made of aluminum); fractional weights 3mg and below are made of aluminum. Weights 500g and smaller are supplied in rugged polycarbonate cases. Sets that include weights 1kg and larger are supplied in oak, aluminum, or rugged plastic cases. All cases are fitted with individual compartments and marked with the accuracy class and serial number. A clear fitted cover secures the fractional weightsMore Product Information