VWR® Dylastir® Magnetic Stirrer
Supplier: VWR International
12620-974
12620-974EA 792.93 CAD
12620-974
VWR® Dylastir® Magnetic Stirrer
Stirrers Magnetic Stirrers
Economical and durable, the Dylastir® magnetic stirrer is perfect for routine mixing applications.
  • Stirs from 100 to 1600 rpm
  • 1500 mL capacity
  • Now with Best-in-Class, Exclusive Five-Year Warranty
The powerful motor and magnet stir up to 1500 mL of water with ease
Optional support plate allows stirrer to be mounted to lab frames or ring stands.
Optional, large cast aluminum top plate offers a stable platform for large vessels. Electronic speed control provides accurate and reproducible results over the entire range. Indicator light illuminatesMore Product Information