VWR® Dyla-Dual™ Hotplate Stirrer, 230 V (Export Only)

Supplier: VWR International
12620-972
12620-972EA 883.76 CAD
12620-972
VWR® Dyla-Dual™ Hotplate Stirrer, 230 V (Export Only)
Stirrers Magnetic Hotplate Stirrers
The economical and durable Dyla-Dual™ hot plate stirrer is ideal for general heating and stirring.

  • Large 16.5 cm (6¹/₂") diameter Cast aluminum top plate
  • Stirs from 100 to 1600 rpm
  • Heats up to 400 °C (752 °F)
  • 1500 ml capacity
  • Now with Best-in-Class, Exclusive five years warranty

A large cast aluminum top plate provides an even heating surface.

Built-in support rod holder with locking knob accepts optional support rod and clamp kit for mounting thermometers and temperature probes. Kit includes 45.7 cm (18") support rod, thermometer extension clamp,More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR