Individual Precision Analytical Weights, Class 3, Troemner
Supplier: Troemner
80858289 80858275 80858260 80858229 80858215 80858200 80858184 80858708 80858669 80858639 80858616 80858577 80858537 80858507 80858486 80858448 80858409 80858374 80858334 80858304 80858711 80858672 80858641 80858619 80858580 80858540 80858509 80858489 80858451 80858412 80858377 80858337 80858306 80858291 80858277 80858262 80858246 80858232 80858217 80858203 80858187 80858709 80858670 80858640 80858617 80858578 80858538 80858508 80858487 80858449 80858410 80858375 80858335 80858290 80858276 80858261 80858231 80858216 80858202 80858186 80858168 80858169 80858141 80858142 80858143 80858127 80858128 80858129 80858114 80858115 80858087 80858088 80858089 80858243 80858305 80858245
12365-326EA 190.91 CAD
12365-326 12365-328 12365-330 12365-334 12365-336 12365-338 12365-340 12765-300 12765-302 12765-304 12765-306 12765-308 12765-310 12765-312 12765-314 12765-316 12765-318 12765-320 12765-322 12765-324 12765-345 12765-346 12765-347 12765-348 12765-349 12765-350 12765-351 12765-352 12765-353 12765-354 12765-355 12765-356 12765-357 12765-358 12765-359 12765-360 12765-361 12765-362 12765-363 12765-364 12765-365 12765-370 12765-371 12765-372 12765-373 12765-374 12765-375 12765-376 12765-377 12765-378 12765-379 12765-380 12765-381 12765-383 12765-384 12765-385 12765-387 12765-388 12765-389 12765-390 14216-754 14216-756 14216-766 14216-768 14216-770 14216-778 14216-780 14216-782 14216-790 14216-792 14216-802 14216-804 14216-806 12365-332 12765-382 12765-386
Individual Precision Analytical Weights, Class 3, Troemner
Weights Calibration Weights
Highly polished kilogram and gram weights made of Troemner Alloy 8, which is a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage
Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are aluminum); fractional weights 3 mg and below are made of aluminum.
This exclusive material has a density of 8.0 at 25 °C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic underMore Product Information