Stainless Steel Calibration Weights, ASTM, Class 1, Ohaus®
Supplier: Ohaus
80780027 80780024 80780021 80780029 80780026 80780023 80780020 80780028 80780022 80780025
11384-120EA 1358.89 CAD
11384-120 11384-122 11384-124 11384-126 11384-128 11384-130 11384-132 11384-134 11384-136 11384-138
Stainless Steel Calibration Weights, ASTM, Class 1, Ohaus®
Weights Calibration Weights
Manufactured to international standards, individual calibration weights are available for a variety of precision applications
Calibrated to ASTM Class 1. Extra-fine accuracy for calibrating high precision analytical balances with a readability as low as 0.1 to 0.01mg.
Order Now
SPECIFICATIONS
Environmentally Preferable