PYREX® Square Bottles, Graduated, Corning
Supplier: Corning
1396-100 1396-250 1396-1L 1396-500 1396-100
11310-680CS 312.63 CAD
11310-680 11310-682 11310-686 11310-684 470211-366
PYREX® Square Bottles, Graduated, Corning
Bottles Glass Bottles
Made of PYREX® borosilicate glass, with linerless, one-piece, orange PP screw cap with a drip-free pouring ring.
  • Exceptional chemical and thermal resistance
  • Rounded edges allow easy cleaning
  • White graduations and a large marking spot
  • Glass ridge moulded into the bottle indicates full capacity
  • Autoclavable
Square design saves 13 to 20% of shelf space, and slightly convex walls minimize glass-to-glass contact during autoclaving.
Further PP colored screw caps for ease of sorting and identification, vented membrane screw caps with 0.22 µm PTFEMore Product Information