Whirl-Pak® Specialty Bags, Nasco Sampling
Supplier: NASCO SAMPLING LLC
B01324 B01319 B01350
11216-550CS 534.54 CAD
11216-550 11216-516 13500-387
Whirl-Pak® Specialty Bags, Nasco Sampling
Bags Sample Bags
Blended, virgin polyethylene film provides exceptional clarity and superior strength HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilization, post-manufacturing sterilization: Increased sample integrity, perforated closure to maintain sterility
  • For use in your meat and poultry facilities
  • Continuous perimeter seal: Maximum protection against leaks
Specially designed products for meat and poultry sampling, Poultry rinse polyethylene bag is sealed at the top to maintain sterility, and has a perforation for easy opening. It will hold smallMore Product Information