VWR® Wafer Box Sealing Tape, PE
Supplier: VWR International
52B-2TR 52B-1RE 52B-1DB 52B-1WH 52B-1BL 52B-1DG 52B-1OR 52B-1YE 52B-1TR 52B-2WH 52B-3WH 52B-1TRSMF 52B-34TR
11211-910CS 967.99 CAD
11211-910 11211-920 11211-922 11211-924 11211-926 11211-928 11211-930 11211-932 11211-934 43300-976 89133-258 89402-996 89501-298
VWR® Wafer Box Sealing Tape, PE
Tapes
Ideal for packaging and transporting of wafer boxes, this low-density polyethylene tape is low in halogens and leachable chlorides.
  • Metal free adhesive
  • Polyethylene film fabrication
  • Acrylic adhesive minimizes outgassing
  • Excellent chemical resistance
  • Good conformability
Polyethylene film and acrylic adhesive minimize outgassing and offer good 'clean peel' properties. Offers excellent chemical resistance and good conformability.
Order Now
SPECIFICATIONS
Environmentally Preferable Waste Reducing