Dissolution Sampling Cannula, Quality Lab Accessories
Supplier: QLA
CA8015-PV CA8015-VK CA8475-PV CA8475-VK CA8775-PV CA8775-VK CA8STP-DK CA8STP-VK CAN013-ST CAN013-ST-5 CAN013-ST-9 CAN015-PH CAN015-SH CAN01K-DK CAN475-01 CAN475-HR CAN475-PH CAN475-SH CAN475-VK CAN500-60 CAN500-DK CAN775-01 CAN775-HR CAN775-PH CAN775-SH CAN775-VK CAN900-45 CANFAE-DK CANFLA-DK CANFLE-DK CANRET-25 CANRET-DK CANS15-PH CANSTP-DK CANSTP-HR CANSTP-ST CANSTP-VK FILASY-DK PRBFSR-ST PRBNSR-ST PROBEX-VK CA9STP-DV CAN500-DSH CAN900-DSH CAN900-DV CANSTP-DVN CA8475-VKS CAN475-01L CAN475-062L CAN475-SHL CAN5BM-DK CAN5BM-DV CAN775-01L CAN775-062L CAN9BM-DK CAN9BM-DV CANS15-SH CANTOP-DK CANTOP-DV PRBFSR-5S PRBFSR-9S CANSTP-06531 CANSTP-06553 CANSTP-07831 CANSTP-07853 CANSTP-12531 CANSTP-12553 CANSTP-14231 CANSTP-14253 CAN015-EW CANSTP-EW CAN02K-DK CAN01K-DV CAN02K-DV CAN500-DV CAN013-ST-5-9 CANSTP08270XDS CANSTP12570XDS STPR-HR THD90-065 CAN01K-DDV CAN01K-DHV CAN01K-DSH CAN01K-DVV CAN500-DDV CAN500-DHV CAN500-DVV CAN900-DDV CAN900-DHV CAN900-DVV ORING-CANDK ORING-CANDV 010-06-202 FILASY-DV PRBFS10-ST PRBFS10-ST82
11010-050EA 264.86 CAD
11010-050 11010-052 11010-054 11010-056 11010-058 11010-060 11010-062 11010-064 11010-070 11010-072 11010-074 11010-078 11010-080 11010-082 11010-084 11010-086 11010-088 11010-090 11010-092 11010-094 11010-096 11010-100 11010-102 11010-104 11010-106 11010-108 11010-110 11010-116 11010-118 11010-120 11010-122 11010-124 11010-126 11010-128 11010-130 11010-132 11010-134 11010-300 11010-432 11010-436 11010-438 89039-788 89039-792 89039-796 89039-798 89039-802 89049-194 89049-204 89049-206 89049-208 89049-210 89049-212 89049-214 89049-216 89049-220 89049-222 89049-228 89049-230 89049-232 89049-304 89049-306 89049-338 89049-340 89049-342 89049-344 89049-346 89049-348 89049-350 89049-352 89132-966 89132-968 93001-032 97002-360 97002-362 97002-364 75848-852 75848-856 75848-858 75848-966 75848-976 75849-020 75849-022 75849-024 75849-026 75849-028 75849-030 75849-032 75849-034 75849-036 75849-038 75849-066 75849-068 75849-086 75849-202 75849-260 75849-262
Dissolution Sampling Cannula, Quality Lab Accessories
Dissolution Apparatus
QLA manufactures a wide variety of Sampling Cannula and Probes for the Dissolution Industry.
  • Manual sampling cannula are manufactured in electroplished 316 Stainless Steel or PEEK materials and come in a variety of lengths
Many industry standard cannula configurations and unique, proprietary QLA configurations are available for most manual and automated sampling applications
The USP defines the sampling point as: "A zone half way between the top of the media and top of the paddle or basket not less than 1 cm from the vessel wall. The samplingMore Product Information