WHEATON® Arsine Generator Apparatus with Standard Taper 24/40 Clear-Seal™ Joint, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
180020 180021 180022 297784-12
10869-000CS 361.5 CAD
10869-000 10869-003 10869-006 80093-776
WHEATON® Arsine Generator Apparatus with Standard Taper 24/40 Clear-Seal™ Joint, DWK Life Sciences
Digestion Systems Trace Element Detection Apparatus
Arsine Generator with standard taper 24/40 Clear-Seal™ Joint engineered for arsine analysis.
  • Arsine analysis done using the silver diethyldithiocarbamate (SDDC) colorimetric method
  • The arsine absorber has a socket joint
  • Made from WHEATON 33 low extractable borosilicate glass
The scrubber tube has a standard taper 24/40 Clear-Seal™ joint and a spherical 12/2 ball joint held together by a size 12 stainless steel ball joint clamp.
Certifications: Meets ASTM/USP and EPA specifications. WHEATON 33 low extractable borosilicate glass conforms to ASTM E 438More Product Information