Tedlar® Sampling Bags, Restek
Supplier: Restek
22050 22053 22049 22057 22051 22055 22054 22058 22052 22056
10058-714PK 385.17 CAD
10058-714 10803-190 10803-272 10862-524 10862-610 10862-764 10862-804 10862-806 10863-026 10863-102
Tedlar® Sampling Bags, Restek
Bags Gas Bags
Sampling bags are a low-cost whole-air sampling device for high-level VOCs and permanent gases.
  • Find the bags you need - we offer sizes from 0.5 to 100 L
  • Unique all-in-one septum and valve fitting make these lightweight and easy to use
  • Polypropylene valve conveniently connects to ³/₁₆" Int.Ø PTFE tubing
  • Continuous sampling temperature up to 107 °C (225 °F); short term (1 to 2 hours) temperature up to 176 °C (350 °F)
Several EPA, NIOSH, and OSHA methods exist for bag sampling for a variety of applications: stationary sources emissions, workplaceMore Product Information