Thermal Desorption Unit (TDU) Tubes (Unconditioned and Conditioned & Capped), Restek
Supplier: Restek
24090 24058 24062 24081 24063 24080 24060 24085 24082 24088 24066 24089 24065 24056 24084 24083 24086 24061
10862-584PK 2252.45 CAD
10862-584 10862-632 10862-634 10862-694 10862-744 10862-766 10862-816 10862-916 10862-918 10862-958 10863-002 10863-004 10863-024 10863-042 10863-066 10863-100 10863-104 10863-110 10863-128 10863-148 10863-152 10863-200 10863-248 10863-258
Thermal Desorption Unit (TDU) Tubes (Unconditioned and Conditioned & Capped), Restek
Chromatography Supplies Chromatography Thermal Desorber Tubes
High-quality thermal desorption tubes are suitable for ppt to ppm concentrations of volatile organic compounds (VOCs) in ambient, indoor, personal, and industrial hygiene environments.
  • Variety of sorbents to collect a wide range of VOCs
  • Use corrosion-resistant glass tubes for excellent inertness
  • Allow monitoring of sorbent bed condition
  • Choose stainless steel tubes for greater durability in the field
  • No sampling pump necessary for passive sampling with diffusion caps
  • Individually etched with unique serial number for convenient sample identification
  • The
More Product Information