VWR® AC Round Bottles, Clear, Wide Mouth
Supplier: VWR International
293090 293058 293068 293073 293071 293091
10861-988CS 407.86 CAD
10861-988 10861-990 10862-226 10862-230 10862-232 10862-238 10862-242 10862-244 10862-246 10862-248 10862-250 10862-254 10862-256 10862-258 10862-356 10862-358 10862-360 10862-362 10862-233 10862-247 10862-255 10862-259
VWR® AC Round Bottles, Clear, Wide Mouth
Bottles Glass Bottles
VWR® Clear, wide mouth AC round bottles are manufactured from soda-lime glass that conforms to USP Type III requirements.
  • Soda-lime glass fabrication
  • No graduations
  • Available in a variety of package sizes with different closure options
These bottles do not feature graduations.
They may be purchased in convenience packs with closures attached to help maintain cleanliness, in bulk packs with unattached closures in separate bags, or in bulk packs without closures.
Packaging: Bottles are available in three packaging options.