VWR® Tissue Culture Flasks, Non-Treated, Sterilized, Non-Pyrogenic
Supplier: VWR International
TCF001025 TCF002025 TCF001050 TCF002050 TCF001250 TCF002250 TCF001600 TCF002600 TCF001850 TCF002850 TCF101600 TCF102600
10861-566CS 472.65 CAD
10861-566 10861-568 10861-570 10861-572 10861-574 10861-576 10861-578 10861-580 10861-582 10861-584 76211-292 76211-294
VWR® Tissue Culture Flasks, Non-Treated, Sterilized, Non-Pyrogenic
Flasks Cell Culture Flasks
Cell and tissue culture flasks are ideal for cell growth and cell yields.
  • Available with five different volume ranges
  • Triangular top with wide base provides stability
  • Protruding ridge on the back side of the flask facilitates stacking
  • Engraved graduations on multiple sides
  • Strict integrity tested
  • Sterilized by electron beam irradiation
  • Non-pyrogenic and not autoclavable
Flasks are designed for use in applications where cell attachment is not desired. They offer an operating temperature range of –20 to +50 °C (–4 to +122 °F). Two different cap stylesMore Product Information