55 Gallon Open-Head UN Rated Steel Drum, New Pig®
Supplier: New Pig
DRM423-LD DRM423-ULD
10860-602EA 917.71 CAD
10860-602 10860-604
55 Gallon Open-Head UN Rated Steel Drum, New Pig®
Barrels, Kegs, Drums
Tough, UN Rated drums are made for hazardous materials; you can rely on them to deliver safety and compliance.
  • UN Rated and tested to help ensure safe shipping of hazardous materials
  • Open-head design and removable lid offer full, convenient access to inside of drum
  • 12-gauge bolt ring closure, sponge rubber gasket and plated bolt/nut provide tight seal to help prevent leaks and spills
  • Interior is coated with a corrosion inhibitor; choose optional phenolic/epoxy lining for added resistance
16-gauge steel lid and 18-gauge steel body and bottom meetMore Product Information